Anti-PA2G4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-706
- 77844-707
- Antigen Symbol:
- PA2G4
- PA2G4
- Antigen Synonyms:
- 38kDa|AA672939|Cell cycle protein p38 2G4 homolog|Cell cycle protein p38-2G4 homolog|ErbB-3 binding protein 1 |ErbB3 binding protein 1 |ErbB3-binding protein 1|ErbB3-binding protein Ebp1 |hG4 1|hG4-1|IRES-specific cellular trans-acting factor 45 kDa|MGC81621|MGC94070|Mpp1|p38 2G4 |Pa2g4|PA2G4_HUMAN|Plfap|Proliferation associated 2G4|38-KD|Proliferation-associated 2G4|38kDa |a|Proliferation-associated protein 1|Proliferation-associated protein 2G4|Protein p38-2G4|si:dz150i12.2|wu:fb19b11|wu:ft56d05|zgc:86732
- 38kDa|AA672939|Cell cycle protein p38 2G4 homolog|Cell cycle protein p38-2G4 homolog|ErbB-3 binding protein 1 |ErbB3 binding protein 1 |ErbB3-binding protein 1|ErbB3-binding protein Ebp1 |hG4 1|hG4-1|IRES-specific cellular trans-acting factor 45 kDa|MGC81621|MGC94070|Mpp1|p38 2G4 |Pa2g4|PA2G4_HUMAN|Plfap|Proliferation associated 2G4|38-KD|Proliferation-associated 2G4|38kDa |a|Proliferation-associated protein 1|Proliferation-associated protein 2G4|Protein p38-2G4|si:dz150i12.2|wu:fb19b11|wu:ft56d05|zgc:86732
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UQ80
- Q9UQ80
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Proliferation-associated protein 2G4 protein (2-394AA)
- Recombinant Human Proliferation-associated protein 2G4 protein (2-394AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice