Anti-EIF4B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77854-620
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:EIF4B
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P23588
- Antigen Synonyms:2310046H11Rik| AL024095| C85189| EIF 4B| eIF-4B| EIF4B| Eukaryotic initiation factor 4B| Eukaryotic translation initiation factor 4B| IF4B_HUMAN| PRO1843
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Eukaryotic translation initiation factor 4B protein (362-590AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: EIF4B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human