Anti-GGA2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: GGA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77844-584
- 77844-583
- Antigen Symbol:
- GGA2
- GGA2
- Antigen Synonyms:
- 1200007E24Rik|ADP ribosylation factor binding protein 2|ADP ribosylation factor binding protein GGA 2|ADP ribosylation factor binding protein GGA2|ADP-ribosylation factor-binding protein GGA2|ARF-binding protein 2|FLJ20966|Gamma adaptin related protein 2|gamma ear-containing|Gamma-adaptin-related protein 2|GGA 2|Gga2|GGA2_HUMAN|Golgi associated gamma adaptin ear containing ARF binding protein 2|Golgi localized gamma ear containing ARF binding protein 2|Golgi-localized|KIAA1080|Vear|VHS domain and ear domain containing protein|VHS domain and ear domain of gamma adaptin|VHS domain and ear domain of gamma-adaptin
- 1200007E24Rik|ADP ribosylation factor binding protein 2|ADP ribosylation factor binding protein GGA 2|ADP ribosylation factor binding protein GGA2|ADP-ribosylation factor-binding protein GGA2|ARF-binding protein 2|FLJ20966|Gamma adaptin related protein 2|gamma ear-containing|Gamma-adaptin-related protein 2|GGA 2|Gga2|GGA2_HUMAN|Golgi associated gamma adaptin ear containing ARF binding protein 2|Golgi localized gamma ear containing ARF binding protein 2|Golgi-localized|KIAA1080|Vear|VHS domain and ear domain containing protein|VHS domain and ear domain of gamma adaptin|VHS domain and ear domain of gamma-adaptin
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UJY4
- Q9UJY4
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human ADP-ribosylation factor-binding protein GGA2 protein (1-300AA)
- Recombinant Human ADP-ribosylation factor-binding protein GGA2 protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice