Anti-Ddt Rabbit Recombinant Polyclonal Antibody
77854-070
New Product
- Antigen Symbol:Ddt
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:O35215
- Antigen Synonyms:DdtD-dopachrome decarboxylase|EC 4.1.1.84|D-dopachrome tautomerase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Mouse D-dopachrome decarboxylase protein (2-118AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: Ddt
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse