Anti-MYRF Rabbit Recombinant Polyclonal Antibody
77846-473
New Product
- Antigen Symbol:MYRF
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y2G1
- Antigen Synonyms:C11orf9|Chromosome 11 open reading frame 9|GM1804|GM98|KIAA0954|MGC10781|MRF|MRF_HUMAN|Myelin gene regulatory factor|Ndt80
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Myelin regulatory factor protein (843-1004AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:20 to 1:200
Type:
Antigen: MYRF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse