Anti-MAFB Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-590
- 77846-591
- Antigen Symbol:
- MAFB
- MAFB
- Antigen Synonyms:
- Kreisler|Kreisler (mouse) maf related leucine zipper homolog|Kreisler maf related leucine zipper homolog|KRML|MAF bZIP transcription factor B|Maf-B|MAFB/Kreisler basic region/leucine zipper transcription factor|MAFB_HUMAN|MGC43127|Segmentation protein KR|Transcription factor V maf musculoaponeurotic fibrosarcoma oncogene homolog B (avian)|V-maf musculoaponeurotic fibrosarcoma oncogene homolog B
- Kreisler|Kreisler (mouse) maf related leucine zipper homolog|Kreisler maf related leucine zipper homolog|KRML|MAF bZIP transcription factor B|Maf-B|MAFB/Kreisler basic region/leucine zipper transcription factor|MAFB_HUMAN|MGC43127|Segmentation protein KR|Transcription factor V maf musculoaponeurotic fibrosarcoma oncogene homolog B (avian)|V-maf musculoaponeurotic fibrosarcoma oncogene homolog B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5Q3
- Q9Y5Q3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Transcription factor MafB protein (168-323AA)
- Recombinant Human Transcription factor MafB protein (168-323AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice