Anti-FZR1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FZR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-173
- 77845-172
- Antigen Symbol:
- FZR1
- FZR1
- Antigen Synonyms:
- CDC20 like 1b|CDC20 like protein 1|CDC20-like protein 1|CDC20C|CDH1|Cdh1/Hct1 homolog|Fizzy related protein 1|Fizzy related protein|Fizzy related protein homolog|Fizzy-related protein homolog|Fizzy/cell division cycle 20 related 1|FYR|FZR 1|FZR 2|Fzr|FZR_HUMAN|Fzr1|Fzr1 protein|FZR2|HCDH 1|HCDH|hCDH1|KIAA1242
- CDC20 like 1b|CDC20 like protein 1|CDC20-like protein 1|CDC20C|CDH1|Cdh1/Hct1 homolog|Fizzy related protein 1|Fizzy related protein|Fizzy related protein homolog|Fizzy-related protein homolog|Fizzy/cell division cycle 20 related 1|FYR|FZR 1|FZR 2|Fzr|FZR_HUMAN|Fzr1|Fzr1 protein|FZR2|HCDH 1|HCDH|hCDH1|KIAA1242
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UM11
- Q9UM11
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Fizzy-related protein homolog protein (1-493AA)
- Recombinant Human Fizzy-related protein homolog protein (1-493AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice