Anti-ADH5 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
77850-788
New Product
- Antigen Symbol:ADH5
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P11766
- Antigen Synonyms:ADH 3|ADHX|ADHX_HUMAN|Alcohol dehydrogenase (class III) chi polypeptide|alcohol dehydrogenase 5 (class III) chi polypeptide|Alcohol dehydrogenase 5|Alcohol dehydrogenase class 3|Alcohol dehydrogenase class chi chain|Alcohol dehydrogenase class III|Alcohol dehydrogenase class-3|Alcohol dehydrogenase class-III|class III alcohol dehydrogenase 5 chi subunit|FALDH|FDH|formaldehyde dehydrogenase|Glutathione dependent formaldehyde dehydrogenase|Glutathione-dependent formaldehyde dehydrogenase|GSH-FDH|GSNOR|hydroxymethyllutathione dehydrogenase|S-(hydroxymethyl)glutathione dehydrogenase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Alcohol dehydrogenase class-3 protein (14-130AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: ADH5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human