Anti-HOXC8 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:1000 to 1:2000, IHC:1:25 to 1:100
Type:
Antigen: HOXC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77848-432
- 77848-433
- Antigen Symbol:
- HOXC8
- HOXC8
- Antigen Synonyms:
- Homeo box 3A|Homeo box 8|Homeobox 3A|Homeobox C8|Homeobox protein Hox C8|Homeobox protein Hox-3A|Homeobox protein Hox-C8|Homeobox protein Homeobox protein R4|Homolog of mouse Hox 3.1|HOX 3|Hox 3.1 mouse homolog of|HOX 3A|HOX C8|Hox-3.1|Hox-3A|HOX3|HOX3A|HOXC 8|Hoxc-8|HXC8_HUMAN|M31
- Homeo box 3A|Homeo box 8|Homeobox 3A|Homeobox C8|Homeobox protein Hox C8|Homeobox protein Hox-3A|Homeobox protein Hox-C8|Homeobox protein Homeobox protein R4|Homolog of mouse Hox 3.1|HOX 3|Hox 3.1 mouse homolog of|HOX 3A|HOX C8|Hox-3.1|Hox-3A|HOX3|HOX3A|HOXC 8|Hoxc-8|HXC8_HUMAN|M31
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31273
- P31273
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human HOXC8
- Synthetic peptide of Human HOXC8
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice