Anti-HEY1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HEY1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-927
- 77845-928
- Antigen Symbol:
- HEY1
- HEY1
- Antigen Synonyms:
- Basic helix loop helix protein OAF1|bHLHb31|Cardiovascular helix loop-helix factor 2|Cardiovascular helix-loop-helix factor 2|CHF-2|CHF2|Class B basic helix-loop-helix protein 31|Hairy and enhancer of split related protein 1|Hairy and enhancer of split-related protein 1|Hairy enhancer-of-split related with YRPW motif protein 1|Hairy related transcription factor 1|Hairy-related transcription factor 1|Hairy/enhancer of split related with YRPW motif 1|Hairy/enhancer-of-split related with YRPW motif protein 1|HERP2|HES related repressor protein 1|HES-related repressor protein 1|HES-related repressor protein 2|HESR-1|HESR1|HEY 1|HEY1_HUMAN|hHRT1|HRT-1|HRT1|OAF1
- Basic helix loop helix protein OAF1|bHLHb31|Cardiovascular helix loop-helix factor 2|Cardiovascular helix-loop-helix factor 2|CHF-2|CHF2|Class B basic helix-loop-helix protein 31|Hairy and enhancer of split related protein 1|Hairy and enhancer of split-related protein 1|Hairy enhancer-of-split related with YRPW motif protein 1|Hairy related transcription factor 1|Hairy-related transcription factor 1|Hairy/enhancer of split related with YRPW motif 1|Hairy/enhancer-of-split related with YRPW motif protein 1|HERP2|HES related repressor protein 1|HES-related repressor protein 1|HES-related repressor protein 2|HESR-1|HESR1|HEY 1|HEY1_HUMAN|hHRT1|HRT-1|HRT1|OAF1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5J3
- Q9Y5J3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Hairy/enhancer-of-split related with YRPW motif protein 1 protein (200-300AA)
- Recombinant Human Hairy/enhancer-of-split related with YRPW motif protein 1 protein (200-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice