Anti-AMPD2 Rabbit Recombinant polyclonal Antibody
Catalog # 77851-112
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AMPD2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q01433
- Antigen Synonyms:Adenosine monophosphate deaminase 2 (isoform L)|Adenosine monophosphate deaminase 2|adenosine monophosphate deaminase 2 isoform L|AMP deaminase 2|AMP deaminase isoform L|AMPD 2|AMPD|AMPD2_HUMAN|RP5-1160K1.5|SPG63
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human AMP deaminase 2 protein (1-129AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: AMPD2
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human