Anti-DPM1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77854-331
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DPM1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O60762
- Antigen Synonyms:CDGIE|dolichol monophosphate mannose synthase|Dolichol phosphate mannose synthase|Dolichol-phosphate mannose synthase|Dolichol-phosphate mannosyltransferase|Dolichyl phosphate beta D mannosyltransferase|dolichyl phosphate mannosyltransferase polypeptide 1|dolichyl phosphate mannosyltransferase polypeptide 1 catalytic subunit|Dolichyl-phosphate beta-D-mannosyltransferase|DPM synthase|DPM1_HUMAN|Mannose P dolichol synthase|Mannose-P-dolichol synthase|MPD synthase|MPDS
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Dolichol-phosphate mannosyltransferase subunit 1 protein (2-260AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: DPM1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human