Anti-CD200R1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, IHC:1:50 to 1:200
Type:
Antigen: CD200R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-845
- 77846-844
- Antigen Symbol:
- CD200R1
- CD200R1
- Antigen Synonyms:
- CD 200R|CD200 cell surface glycoprotein receptor|CD200 receptor 1|Cell surface glycoprotein CD200 receptor 1|Cell surface glycoprotein OX2 receptor 1|Cell surface glycoprotein OX2 receptor|cell surface glycoprotein receptor CD200|CRTR 2|CRTR2|HCRTR2|MO2R1_HUMAN|MOX2 receptor|MOX2R|OX2R
- CD 200R|CD200 cell surface glycoprotein receptor|CD200 receptor 1|Cell surface glycoprotein CD200 receptor 1|Cell surface glycoprotein OX2 receptor 1|Cell surface glycoprotein OX2 receptor|cell surface glycoprotein receptor CD200|CRTR 2|CRTR2|HCRTR2|MO2R1_HUMAN|MOX2 receptor|MOX2R|OX2R
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TD46
- Q8TD46
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CD200R1
- Fusion protein of Human CD200R1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice