Anti-BIN1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-880
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BIN1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O00499
- Antigen Synonyms:AMPH 2|AMPH2|Amphiphysin 2|Amphiphysin II|Amphiphysin like protein|amphiphysin-like|Amphiphysin-like protein|AMPHL|BIN1_HUMAN|Box Dependant MYC Interacting Protein 1|Box-dependent myc-interacting protein 1|Bridging integrator 1|DKFZp547F068|MGC10367|MGC105358|Myc box dependent interacting protein 1|Myc box-dependent-interacting protein 1|SH3P9
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Myc box-dependent-interacting protein 1 protein (414-593AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: BIN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human