Anti-DCTN5 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: DCTN5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77847-467
- 77847-468
- Antigen Symbol:
- DCTN5
- DCTN5
- Antigen Synonyms:
- DCTN 5|DCTN5_HUMAN|Dynactin 4|Dynactin 5 (p25)|Dynactin subunit 5|Dynactin subunit p25|p25
- DCTN 5|DCTN5_HUMAN|Dynactin 4|Dynactin 5 (p25)|Dynactin subunit 5|Dynactin subunit p25|p25
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BTE1
- Q9BTE1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human DCTN5
- Fusion protein of Human DCTN5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice