Human Creatine Phosphokinase-MM Isoenzyme (from Skeletal muscle)
Catalog # 80051-270
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Source:Skeletal muscle
- Species:Human
- Size:50 IU
- Storage Conditions:Safe to freeze
- Enzyme Name:Creatine Phosphokinase-MM Isoenzyme
- Enzyme Synonyms:hCPK-MM