Anti-FZD4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: FZD4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-943
- 77843-944
- Antigen Symbol:
- FZD4
- FZD4
- Antigen Synonyms:
- CD 344| CD 344 antigen| CD344| CD344 antigen| EVR1| FEVR| Frizzled (Drosophila) homolog 4| Frizzled 4| Frizzled family receptor 4| Frizzled homolog 4 (Drosophila)| Frizzled-4| Fz 4| Fz-4| Fz4| FZD4| FZD4_HUMAN| FZD4S| FzE4| GPCR| hFz4| MGC34390| WNT receptor frizzled 4
- CD 344| CD 344 antigen| CD344| CD344 antigen| EVR1| FEVR| Frizzled (Drosophila) homolog 4| Frizzled 4| Frizzled family receptor 4| Frizzled homolog 4 (Drosophila)| Frizzled-4| Fz 4| Fz-4| Fz4| FZD4| FZD4_HUMAN| FZD4S| FzE4| GPCR| hFz4| MGC34390| WNT receptor frizzled 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULV1
- Q9ULV1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Frizzled-4 protein (37-222AA)
- Recombinant Human Frizzled-4 protein (37-222AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice