Human Recombinant 5-Lipoxygenase (from Spodoptera frugiperda)
Catalog # 80512-114
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Enzyme type:Recombinant
- Source:Spodoptera frugiperda
- Species:Human
- Size:500 U
- Storage Conditions:Safe to freeze, avoid repeated freeze/thaw cycles
- Enzyme Name:5-Lipoxygenase
- Enzyme Synonyms:5-LO