Anti-Alb Rabbit Polyclonal Antibody
Catalog # 77850-463
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Alb
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:P07724
- Antigen Synonyms:Alb 1|ALBU_MOUSE|Albumin 1|Albumin|Serum albumin
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Serum Albumin purified from Mouse serum (Native Protein)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human