Anti-PCDHA12 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PCDHA12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-323
- 77844-324
- Antigen Symbol:
- PCDHA12
- PCDHA12
- Antigen Synonyms:
- KIAA0345 like 2|MGC138485|MGC141932|PCDAC_HUMAN|PCDH alpha 12|PCDH alpha12|PCDH-alpha-12|Protocadherin alpha 12|Protocadherin alpha-12
- KIAA0345 like 2|MGC138485|MGC141932|PCDAC_HUMAN|PCDH alpha 12|PCDH alpha12|PCDH-alpha-12|Protocadherin alpha 12|Protocadherin alpha-12
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UN75
- Q9UN75
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Protocadherin alpha-12 protein (862-941AA)
- Recombinant human Protocadherin alpha-12 protein (862-941AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice