Anti-ATP2C1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-581
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP2C1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:P98194
- Antigen Synonyms:1700121J11Rik|AT2C1_HUMAN|ATP dependent Ca(2+) pump PMR1|ATP-dependent Ca(2+) pump PMR1|ATP2C1A|ATPase 2C1|ATPase Ca(2+) sequestering|ATPase Ca++ transporting type 2C member 1|AW061228|BCPM|Calcium transporting ATPase type 2C member 1|Calcium-transporting ATPase type 2C member 1|D930003G21Rik|HHD|hSPCA1|HUSSY 28|KIAA1347|MGC58010|MGC93231|OTTHUMP00000216066|OTTHUMP00000216068|OTTHUMP00000216069|OTTHUMP00000216071|OTTHUMP00000216072|OTTHUMP00000216073|OTTHUMP00000216074|OTTHUMP00000216075|PMR1|rat|homolog of|PMR1L|Secretory pathway Ca(2+) ATPase 1|Secretory pathway Ca(2+)-transporting ATPase|Secretory pathway Ca2+/Mn2+ ATPase 1|SPCA|SPCA1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Calcium-transporting ATPase type 2C member 1 protein (491-605AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: ATP2C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human