Anti-EEF1D Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77854-509
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:EEF1D
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P29692
- Antigen Synonyms:Antigen NY CO 4|Antigen NY-CO-4|EEF 1D|EEF1D|EF 1 delta|EF 1D|EF-1-delta|EF1D|EF1D_HUMAN|Elongation factor 1 delta|Elongation factor 1-delta|eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein)|Eukaryotic translation elongation factor 1 delta|FP1047|Guanine nucleotide exchange protein
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Elongation factor 1-delta protein (2-281AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: EEF1D
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human