Anti-DGKG Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-135
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DGKG
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P49619
- Antigen Synonyms:DGKG|DAGK3Diacylglycerol kinase gamma|DAG kinase gamma|EC 2.7.1.107|Diglyceride kinase gamma|DGK-gamma
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Diacylglycerol kinase gamma protein (1-255AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: DGKG
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse