Anti-CYSLTR1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
77853-961
New Product
- Antigen Symbol:OR6Q1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8NGQ2
- Antigen Synonyms:OR6Q1|Olfactory receptor 6Q1|Olfactory receptor OR11-226
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Olfactory receptor OR11-226
- Purification:Antigen affinity purified
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human