Anti-ALDH6A1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, WB:1:500 to 1:2000, IHC:1:50 to 1:200
Type:
Antigen: ALDH6A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77847-639
- 77847-638
- Antigen Symbol:
- ALDH6A1
- ALDH6A1
- Antigen Synonyms:
- 1110038I05Rik|AI314632|Aldehyde dehydrogenase 6 family member A1|Aldehyde dehydrogenase family 6 member A1|Aldehyde dehydrogenase family 6, subfamily A1|Malonate semialdehyde dehydrogenase|Malonate-semialdehyde dehydrogenase [acylating]|Methylmalonate semialdehyde dehydrogenase [acylating] mitochondrial|Methylmalonate-semialdehyde dehydrogenase [acylating]|MGC37325|MGC40271|Mitochondrial acylating methylmalonate semialdehyde dehydrogenase|mitochondrial|MMSA_HUMAN|MMSADHA|MMSDH
- 1110038I05Rik|AI314632|Aldehyde dehydrogenase 6 family member A1|Aldehyde dehydrogenase family 6 member A1|Aldehyde dehydrogenase family 6, subfamily A1|Malonate semialdehyde dehydrogenase|Malonate-semialdehyde dehydrogenase [acylating]|Methylmalonate semialdehyde dehydrogenase [acylating] mitochondrial|Methylmalonate-semialdehyde dehydrogenase [acylating]|MGC37325|MGC40271|Mitochondrial acylating methylmalonate semialdehyde dehydrogenase|mitochondrial|MMSA_HUMAN|MMSADHA|MMSDH
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02252
- Q02252
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ALDH6A1
- Fusion protein of Human ALDH6A1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice