Anti-DCT Rabbit Polyclonal Antibody
Catalog # 77847-979
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DCT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P40126
- Antigen Synonyms:L dopachrome tautomerase|Dopachrome delta isomerase|Dopachrome tautomerase|Dopachrome tautomerase dopachrome delta isomerase|tyrosine related protein 2|DT|EC 5.3.3.12|L dopachrome Delta isomerase|L-dopachrome Delta-isomerase|L-dopachrome tautomerase|TRP 2|TRP-2|TRP2|Tyrosinase related protein 2|Tyrosinase-related protein 2|TYRP2|TYRP2_HUMAN
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human DCT
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000, IF: 1:100 to 1:500
Type:
Antigen: DCT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse