Anti-KMT2B Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: KMT2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77844-112
- 77844-111
- Antigen Symbol:
- KMT2B
- KMT2B
- Antigen Synonyms:
- Histone-lysine N-methyltransferase 2B|HRX2|KIAA0304 gene product|KMT2B_HUMAN|Likely ortholog of mouse WW domain binding protein 7|Lysine N-methyltransferase 2B|mixed lineage leukemia 4|Mixed lineage leukemia gene homolog 2 protein|MLL2|myeloid lymphoid leukemia 4|Myeloid lymphoid or mixed lineage leukemia protein 4|Myeloid/lymphoid or mixed-lineage leukemia protein 4|Trithorax homolog 2|trithorax homologue 2|TRX2|WBP-7|WBP7|WW domain-binding protein 7
- Histone-lysine N-methyltransferase 2B|HRX2|KIAA0304 gene product|KMT2B_HUMAN|Likely ortholog of mouse WW domain binding protein 7|Lysine N-methyltransferase 2B|mixed lineage leukemia 4|Mixed lineage leukemia gene homolog 2 protein|MLL2|myeloid lymphoid leukemia 4|Myeloid lymphoid or mixed lineage leukemia protein 4|Myeloid/lymphoid or mixed-lineage leukemia protein 4|Trithorax homolog 2|trithorax homologue 2|TRX2|WBP-7|WBP7|WW domain-binding protein 7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UMN6
- Q9UMN6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Histone-lysine N-methyltransferase 2B protein (145-290AA)
- Recombinant Human Histone-lysine N-methyltransferase 2B protein (145-290AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice