Anti-DCAF1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: DCAF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-682
- 77845-683
- Antigen Symbol:
- DCAF1
- DCAF1
- Antigen Synonyms:
- DCAF1|KIAA0800|RIP|VPRBPDDB1- and CUL4-associated factor 1|HIV-1 Vpr-binding protein|VprBP|Serine/threonine-protein kinase VPRBP|EC 2.7.11.1|Vpr-interacting protein
- DCAF1|KIAA0800|RIP|VPRBPDDB1- and CUL4-associated factor 1|HIV-1 Vpr-binding protein|VprBP|Serine/threonine-protein kinase VPRBP|EC 2.7.11.1|Vpr-interacting protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4B6
- Q9Y4B6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DDB1- and CUL4-associated factor 1 protein (647-745AA)
- Recombinant Human DDB1- and CUL4-associated factor 1 protein (647-745AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice