Anti-WNT9A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: WNT9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77848-627
- 77848-628
- Antigen Symbol:
- WNT9A
- WNT9A
- Antigen Synonyms:
- MGC138165|MGC141991|Protein Wnt-14|Protein Wnt-9a|RP23-378H5.1|Wingless related MMTV integration site 9A|Wingless type MMTV integration site 9A|Wingless type MMTV integration site family|member 9A|wingless-type MMTV integration site family|member 14|Wnt 14|Wnt 9a|WNT14|WNT9A|WNT9A_HUMAN
- MGC138165|MGC141991|Protein Wnt-14|Protein Wnt-9a|RP23-378H5.1|Wingless related MMTV integration site 9A|Wingless type MMTV integration site 9A|Wingless type MMTV integration site family|member 9A|wingless-type MMTV integration site family|member 14|Wnt 14|Wnt 9a|WNT14|WNT9A|WNT9A_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14904
- O14904
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human WNT9A
- Fusion protein of Human WNT9A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice