Anti-ASCC1 Rabbit Recombinant Polyclonal Antibody
77851-460
New Product
- Antigen Symbol:ASCC1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8N9N2
- Antigen Synonyms:Activating signal cointegrator 1 complex subunit 1|ASC-1 complex subunit p50|ASC1 complex subunit p50|ASC1p50|ASCC1_HUMAN|CGI 18|RP11-150D20.4|Trip4 complex subunit p50
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Activating signal cointegrator 1 complex subunit 1 protein (1-315AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ASCC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse