Anti-CRYBA4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77853-609
New Product
- Antigen Symbol:CRYBA4
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P53673
- Antigen Synonyms:Beta A4 crystallin|Beta crystallin A4|Beta-A4 crystallin|Beta-crystallin A4|CRBA4_HUMAN|CRYBA4|Crystallin beta A4|Crystallin|beta polypeptide A4|CTRCT23|Eye lens structural protein|MCOPCT4
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-crystallin A4 protein (2-196AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: CRYBA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human