Anti-CHAT Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77853-044
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CHAT
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P28329
- Antigen Synonyms:Acetyl CoA choline O acetyltransferase|Acetyl CoA:choline O acetyltransferase|CHOACTase|Choline acetylase|choline acetyltransferase|Choline O acetyltransferase|Choline O-acetyltransferase|CLAT_HUMAN|CMS1A|CMS1A2|EC 2.3.1.6|OTTHUMP00000019583|OTTHUMP00000019584
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Choline O-acetyltransferase protein (354-592AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CHAT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human