Anti-BOK Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:2000 to 1:5000, IHC:1:25 to 1:100
Type:
Antigen: BOK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77846-897
- 77846-898
- Antigen Symbol:
- BOK
- BOK
- Antigen Synonyms:
- Bcl 2 related ovarian killer protein|Bcl-2-like protein 9|Bcl-2-related ovarian killer protein|BCL2 related ovarian killer|Bcl2-L-9|BCL2L9|BOK_HUMAN|BOKL|HMGC4631
- Bcl 2 related ovarian killer protein|Bcl-2-like protein 9|Bcl-2-related ovarian killer protein|BCL2 related ovarian killer|Bcl2-L-9|BCL2L9|BOK_HUMAN|BOKL|HMGC4631
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UMX3
- Q9UMX3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human BOK(BH3 domain)
- Synthetic peptide of Human BOK(BH3 domain)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice