Anti-CCR10 Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-548
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CCR10
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P46092
- Antigen Synonyms:C C chemokine receptor type 10|C-C chemokine receptor type 10|C-C CKR-10|CC chemokine receptor 10|CC CKR 10|CC-CKR-10|CCR-10|CCR10_HUMAN|Chemokine (C C motif) receptor 10|G protein coupled receptor 2|G-protein coupled receptor 2|GPR2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human C-C chemokine receptor type 10 protein (314-362AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:200 to 1:5000, IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: CCR10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human