Anti-TACC3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: TACC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77846-006
- 77846-005
- Antigen Symbol:
- TACC3
- TACC3
- Antigen Synonyms:
- ERIC 1|ERIC-1|ERIC1|MGC117382|MGC133242|OTTHUMP00000113796|TACC3_HUMAN|Transforming acidic coiled coil containing protein 3|Transforming acidic coiled-coil-containing protein 3
- ERIC 1|ERIC-1|ERIC1|MGC117382|MGC133242|OTTHUMP00000113796|TACC3_HUMAN|Transforming acidic coiled coil containing protein 3|Transforming acidic coiled-coil-containing protein 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6A5
- Q9Y6A5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Transforming acidic coiled-coil-containing protein 3 protein (133-268AA)
- Recombinant Human Transforming acidic coiled-coil-containing protein 3 protein (133-268AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice