Anti-RTRAF Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RTRAF
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-541
- 77845-542
- Antigen Symbol:
- RTRAF
- RTRAF
- Antigen Synonyms:
- C14orf166|CGI 99|CGI-99|CGI99|Chromosome 14 open reading frame 166|CLE|CLE7|CN166_HUMAN|Homeobox prox 1|LCRP369|RLL motif containing 1|RLLM1|UPF0568 protein C14orf166
- C14orf166|CGI 99|CGI-99|CGI99|Chromosome 14 open reading frame 166|CLE|CLE7|CN166_HUMAN|Homeobox prox 1|LCRP369|RLL motif containing 1|RLLM1|UPF0568 protein C14orf166
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y224
- Q9Y224
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human RNA transcription, translation and transport factor protein (1-244AA)
- Recombinant Human RNA transcription, translation and transport factor protein (1-244AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice