Anti-HOXA2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77857-559
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HOXA2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O43364
- Antigen Synonyms:AI324701|homeo box A2|homeobox A2|Homeobox protein Hox-1K|Homeobox protein Hox-A2|Hox-1.11|HOX1.11|HOX1K|Hoxa-2|Hoxa11|HXA2_HUMAN|MGC151482|MGC151484|RATHOX111A
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Homeobox protein Hox-A2 protein (26-124AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:2000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: HOXA2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse