Anti-EIF4EBP2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: EIF4EBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77848-181
- 77848-182
- Antigen Symbol:
- EIF4EBP2
- EIF4EBP2
- Antigen Synonyms:
- 4E-BP2|4EBP2|4EBP2_HUMAN|eIF4E binding protein 2|eIF4E-binding protein 2|Eukaryotic translation initiation factor 4E binding protein 1|Eukaryotic translation initiation factor 4E-binding protein 2|PHASII|phosphorylated|heat and acid stable regulated by insulin protein II
- 4E-BP2|4EBP2|4EBP2_HUMAN|eIF4E binding protein 2|eIF4E-binding protein 2|Eukaryotic translation initiation factor 4E binding protein 1|Eukaryotic translation initiation factor 4E-binding protein 2|PHASII|phosphorylated|heat and acid stable regulated by insulin protein II
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13542
- Q13542
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human EIF4EBP2
- Fusion protein of Human EIF4EBP2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together