Anti-ATP6V1C1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77851-645
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ATP6V1C1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P21283
- Antigen Synonyms:ATP6C|ATP6D|ATP6V1C1|ATPase H+ transporting lysosomal (vacuolar proton pump) 42kD|ATPase H+ transporting lysosomal 42kD V1 subunit C isoform 1|ATPase H+ transporting lysosomal 42kDa V1 subunit C isoform 1|ATPase H+ transporting lysosomal 42kDa V1 subunit C1|ATPase H+ transporting lysosomal V1 subunit C1|FLJ20057|H(+) transporting two sector ATPase subunit C|H+ ATPase C subunit|H+ transporting ATPase chain C vacuolar|Subunit C of vacuolar proton ATPase V1 domain|V ATPase C subunit|V ATPase subunit C 1|V-ATPase subunit C 1|V-type proton ATPase subunit C 1|Vacuolar ATP synthase subunit C|Vacuolar proton pump 42 kD subunit|Vacuolar proton pump C subunit|Vacuolar proton pump subunit C 1|Vacuolar protonATPase subunit C VI domain|VATC|VATC1_HUMAN|VATPase C subunit|VATPase subunit C 1|VMA5
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human V-type proton ATPase subunit C 1 protein (129-382AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ATP6V1C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human