Anti-DLX1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77854-209
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DLX1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P56177
- Antigen Synonyms:DII B|Distal less homeo box 1|Distal less homeobox 1|Distal-less homeobox gene 1a|Dlx 1|Dlx|Dlx1|Dlx1 distal-less homeobox 1|DLX1_HUMAN|Homeo box protein DLX1|Homeobox protein DLX 1|Homeobox protein DLX-1|Homeobox protein Dlx1a|Homeodomain transcription factor DLX1|MGC189409|OTTHUMP00000082494|OTTHUMP00000082497|OTTMUSP00000014202|RP23-419H3.3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Homeobox protein DLX-1 protein (8-114AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: DLX1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human