Anti-ACOX3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77849-342
- 77849-343
- Antigen Symbol:
- ACOX3
- ACOX3
- Antigen Synonyms:
- ACOX3|BRCOX|PRCOXPeroxisomal acyl-coenzyme A oxidase 3|EC 1.3.3.6|Branched-chain acyl-CoA oxidase|BRCACox|Pristanoyl-CoA oxidase
- ACOX3|BRCOX|PRCOXPeroxisomal acyl-coenzyme A oxidase 3|EC 1.3.3.6|Branched-chain acyl-CoA oxidase|BRCACox|Pristanoyl-CoA oxidase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15254
- O15254
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ACOX3
- Fusion protein of Human ACOX3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice