Anti-CHN2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
77853-089
New Product
- Antigen Symbol:CHN2
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P52757
- Antigen Synonyms:ARHGAP3|BCH|Beta-chimaerin|Beta-chimerin|Rho GTPase-activating protein 3
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-chimaerin protein (1-91AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: CHN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human