Anti-CIAO1 Rabbit Recombinant Polyclonal Antibody
77853-157
New Product
- Antigen Symbol:CIAO1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O76071
- Antigen Synonyms:CIA1|WDR39Probable cytosolic iron-sulfur protein assembly protein WD repeat-containing protein 39
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Probable cytosolic iron-sulfur protein assembly protein CIAO1 protein (1-339AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CIAO1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human