Order Entry
Puerto Rico
ContactUsLinkComponent
Human MMP-9 (from Neutrophil)
Human MMP-9 (from Neutrophil)
Catalog # 80053-078
Supplier:  MilliporeSigma
CAS Number:  
Human MMP-9 (from Neutrophil)
Catalog # 80053-078
Supplier:  MilliporeSigma
Supplier Number:  444231-5UG
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Source:
    Neutrophil
  • Species:
    Human
  • Size:
    5 μg
  • Storage Conditions:
    Safe to freeze, avoid repeated freeze/thaw cycles
  • Enzyme Name:
    MMP-9
  • Enzyme Synonyms:
    Matrix Metalloproteinase 9;Gelatinase B;92 kDa Type IV Collagenase
  • Purity:
    ≥90% (SDS-PAGE)

Specifications

About this item