Anti-MBIP Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:200 to 1:500
Type:
Antigen: MBIP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-466
- 77843-465
- Antigen Symbol:
- MBIP
- MBIP
- Antigen Synonyms:
- MAP3K12 binding inhibitory protein 1|MAP3K12-binding inhibitory protein 1|MAPK upstream kinase binding inhibitory protein|MAPK upstream kinase-binding inhibitory protein|Mbip|MBIP1_HUMAN|MUK binding inhibitory protein|MUK-binding inhibitory protein|OTTHUMP00000178844
- MAP3K12 binding inhibitory protein 1|MAP3K12-binding inhibitory protein 1|MAPK upstream kinase binding inhibitory protein|MAPK upstream kinase-binding inhibitory protein|Mbip|MBIP1_HUMAN|MUK binding inhibitory protein|MUK-binding inhibitory protein|OTTHUMP00000178844
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NS73
- Q9NS73
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human MAP3K12-binding inhibitory protein 1 protein (91-241AA)
- Recombinant Human MAP3K12-binding inhibitory protein 1 protein (91-241AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice