Anti-KMT5A Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KMT5A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-405
- 77843-404
- Antigen Symbol:
- KMT5A
- KMT5A
- Antigen Synonyms:
- H4 K20 HMTase|H4 K20 specific histone methyltransferase|H4-K20-HMTase SETD8|Histone H4 K20 methyltransferase|Histone lysine N methyltransferase H4 lysine 20 specific|Histone-lysine N-methyltransferase SETD8|Lysine N-methyltransferase 5A|N-lysine methyltransferase SETD8|Pr SET 7|Pr SET7|PR-Set7|PR/SET domain containing protein 07|PR/SET domain containing protein 8|PR/SET domain-containing protein 07|PR/SET07|PrSET7|SET 07|SET 8|SET domain containing (lysine methyltransferase) 8|SET domain containing 8|SET domain containing lysine methyltransferase 8|SET domain containing protein 8|SET domain-containing protein 8|SET07|SET8|SETD 8|setd8|SETD8_HUMAN
- H4 K20 HMTase|H4 K20 specific histone methyltransferase|H4-K20-HMTase SETD8|Histone H4 K20 methyltransferase|Histone lysine N methyltransferase H4 lysine 20 specific|Histone-lysine N-methyltransferase SETD8|Lysine N-methyltransferase 5A|N-lysine methyltransferase SETD8|Pr SET 7|Pr SET7|PR-Set7|PR/SET domain containing protein 07|PR/SET domain containing protein 8|PR/SET domain-containing protein 07|PR/SET07|PrSET7|SET 07|SET 8|SET domain containing (lysine methyltransferase) 8|SET domain containing 8|SET domain containing lysine methyltransferase 8|SET domain containing protein 8|SET domain-containing protein 8|SET07|SET8|SETD 8|setd8|SETD8_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQR1
- Q9NQR1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human N-lysine methyltransferase KMT5A protein (136-258AA)
- Recombinant human N-lysine methyltransferase KMT5A protein (136-258AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice