Anti-DDO Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77854-068
New Product
- Antigen Symbol:DDO
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q99489
- Antigen Synonyms:D aspartate oxidase|D-aspartate oxidase|DASOX|DDO 1|DDO 2|FLJ45203|OTTHUMP00000017000|OXDD_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human D-aspartate oxidase protein (37-161AA)
- Purification:>95%, protein G purified
- Packaging:Vial
Type:
Antigen: DDO
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human