Anti-TMSB10 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:30 to 1:150
Type:
Antigen: TMSB10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77848-077
- 77848-078
- Antigen Symbol:
- TMSB10
- TMSB10
- Antigen Synonyms:
- MIG 12|MIG12|Migration inducing gene 12|Migration inducing protein 12|Ptmb10|TB 10|TB10|Thyb 10|Thyb10|Thymosin beta-10|TYB10_HUMAN
- MIG 12|MIG12|Migration inducing gene 12|Migration inducing protein 12|Ptmb10|TB 10|TB10|Thyb 10|Thyb10|Thymosin beta-10|TYB10_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P63313
- P63313
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human TMSB10
- Synthetic peptide of Human TMSB10
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice