Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-APOBEC1 Rabbit Recombinant Polyclonal Antibody
Anti-APOBEC1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-290
Supplier:  AFG Bioscience
New Product
undefined
Anti-APOBEC1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-290
Supplier:  AFG Bioscience
Supplier Number:  A12889-50UG
New Product

Specifications

  • Antigen Symbol:
    APOBEC1
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    50 µg
  • Form:
    Liquid
  • Gene ID:
    P41238
  • Antigen Synonyms:
    ABEC1_HUMAN| APOBEC 1| APOBEC1| Apolipoprotein B mRNA editing enzyme| Apolipoprotein B mRNA editing enzyme catalytic polypeptide 1| Apolipoprotein B mRNA editing enzyme complex 1| Apolipoprotein B mRNA-editing enzyme 1| BEDP| C->U-editing enzyme APOBEC-1| CDAR1| EC 3.5.4.| HEPR
  • Storage Buffer:
    0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human C->U-editing enzyme APOBEC-1 protein (1-172AA)
  • Purification:
    >95%, Protein G purified
  • Packaging:
    Vial

Specifications

About this item

Recommended dilution: WB:1:1000 to 1:5000

Type:
Antigen: APOBEC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human