Anti-DAXX HEK293F cells Recombinant Monoclonal Antibody [clone: 3A2]
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DAXX
Clonality: Recombinant Monoclonal
Clone: 3A2
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77850-399
- 77850-398
- Antigen Symbol:
- DAXX
- DAXX
- Antigen Synonyms:
- Death domain-associated protein 6, Daxx, hDaxx, ETS1-associated protein 1, EAP1, Fas death domain-associated protein, DAXX, BING2, DAP6
- Death domain-associated protein 6, Daxx, hDaxx, ETS1-associated protein 1, EAP1, Fas death domain-associated protein, DAXX, BING2, DAP6
- Clonality:
- Recombinant Monoclonal
- Recombinant Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 3A2
- 3A2
- Reactivity:
- Human
- Human
- Host:
- HEK293F cells
- HEK293F cells
- Gene ID:
- Q9UER7
- Q9UER7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- A synthesized peptide derived from Human DAXX
- A synthesized peptide derived from Human DAXX
- Form:
- Liquid
- Liquid
- Purification:
- Affinity purified
- Affinity purified
- Storage Buffer:
- PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice